Learn More
New England Biolabs, Inc. BsaI-HF®v2 – 1000 units
SDP
Catalog No. 501527865
Encompass
BsaI-HFv2 is a Type IIS restriction enzyme that has been optimized for Golden Gate Assembly and in protocols requiring DNA cutting by BsaI. Works well for any protocol requiring DNA cutting by BsaI. Recommended enzyme for any purpose requiring digestion at the recognition sequence ...5-GGTCTC(N1)/(N5)-3...
  • Engineered for improved performance
  • Used in Golden Gate Assembly; maintains high activity levels in T4 DNA Ligase Buffer
  • Type IIS REs recognize asymmetric DNA sequences; cleave outside their recognition sequence
  • 100% activity in rCutSmart Buffer
  • Time-Saver qualified for digestion in 5-15 minutes
Catalog No. 50-152-7865 Supplier New England Biolabs, Inc. Supplier No. R3733S
BsaI-HFv2 is a Type IIS restriction enzyme that has been optimized for Golden Gate Assembly and in protocols requiring DNA cutting by BsaI. Works well for any protocol requiring DNA cutting by BsaI. Recommended enzyme for any purpose requiring digestion at the recognition sequence ...5-GGTCTC(N1)/(N5)-3...
  • Engineered for improved performance
  • Used in Golden Gate Assembly; maintains high activity levels in T4 DNA Ligase Buffer
  • Type IIS REs recognize asymmetric DNA sequences; cleave outside their recognition sequence
  • 100% activity in rCutSmart Buffer
  • Time-Saver qualified for digestion in 5-15 minutes