Learn More
New England Biolabs, Inc. BsaWI – 250 units
SDP
Catalog No. 50812257
Encompass
A restriction endonuclease that recognizes the sequence W^CCGG_W.
Catalog No. 50-812-257 Supplier New England Biolabs, Inc. Supplier No. R0567S
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence W^CCGG_W.