Learn More
New England Biolabs, Inc. BsaXI – 100 units
SDP
Catalog No. 50812340
Encompass
A restriction endonuclease that recognizes the sequence _NNN^NNNNNNNNNACNNNNNCTCCNNNNNNN_NNN^.
Catalog No. 50-812-340 Supplier New England Biolabs, Inc. Supplier No. R0609S
A restriction endonuclease that recognizes the sequence _NNN^NNNNNNNNNACNNNNNCTCCNNNNNNN_NNN^.