Learn More
New England Biolabs, Inc. BsaXI – 500 units
SDP
Catalog No. 50812339
Encompass
A restriction endonuclease that recognizes the sequence _NNN^NNNNNNNNNACNNNNNCTCCNNNNNNN_NNN^.
Catalog No. 50-812-339 Supplier New England Biolabs, Inc. Supplier No. R0609L
A restriction endonuclease that recognizes the sequence _NNN^NNNNNNNNNACNNNNNCTCCNNNNNNN_NNN^.