Learn More
ATCC BS-C-1; Kidney; African green monkey (Chlorocebus aethiops) Grivet
Supplier:  ATCC CCL26
Encompass
BS-C-1 is a cell line exhibiting epithelial morphology isolated from the kidney of an African green monkey, suitable for transfection by SV40 vectors. This cell line was deposited by HE Hopps.
Catalog No. 50-238-4060