Learn More
Alkali Scientific Alkali Scientific™ BSC1006 Mini-Centrifuge, Includes Round Rotor (6 Place), Blue
SDP
Catalog No. 503029664
Encompass_Preferred
The Alkali Scientific™ BSC1006 Mini-Centrifuges are ideal for quick spin-downs of micro samples. The easy-open lid permits convenient single-handed operation. The clear lid and electronic safety cut-off allow safe sample observation. The centrifuge quickly accelerates to maximum speed (6,600 rpm/2,200 rcf) and stops within seconds. The advanced motor mounting system provides quiet, vibration-free operation. The BSC1006 centrifuge includes a traditional round angle rotor (for six standard 1.5ml to 2.0ml microtubes) as well as a strip tube rotor for 2 PCR strips or 16 individual 0.2 ml tubes. Additional adapters are also included for 0.4-0.5ml tubes. The rotors can be easily interchanged within a matter of seconds. The BSC1006 Mini Centrifuge comes in a red  or blue color.
Catalog No. 50-302-9664 Supplier Alkali Scientific Supplier No. RS1006B
The Alkali Scientific™ BSC1006 Mini-Centrifuges are ideal for quick spin-downs of micro samples. The easy-open lid permits convenient single-handed operation. The clear lid and electronic safety cut-off allow safe sample observation. The centrifuge quickly accelerates to maximum speed (6,600 rpm/2,200 rcf) and stops within seconds. The advanced motor mounting system provides quiet, vibration-free operation. The BSC1006 centrifuge includes a traditional round angle rotor (for six standard 1.5ml to 2.0ml microtubes) as well as a strip tube rotor for 2 PCR strips or 16 individual 0.2 ml tubes. Additional adapters are also included for 0.4-0.5ml tubes. The rotors can be easily interchanged within a matter of seconds. The BSC1006 Mini Centrifuge comes in a red  or blue color.