Learn More
Aviva Systems Biology Corp BSCL2 Antibody - C-terminal region (ARP50102_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50102_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BSCL2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human BSCL2. Alternate names: GNG3LG, HMN5, MGC4694, SPG17ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-487