Learn More
Enzo Life Sciences BSEP (human) monoclonal antibody (USal-hBSEP-McAb-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX801035C100
Encompass_Preferred
Clone: USal-hBSEP-McAb-1. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 705-718 (Y705LVHEPPLAVVDHK718) of human BSEP (bile salt export pump). Species reactivity: Human. Applications: ICC, WB. Purity: Protein G-affinity purified. Formulation: Liquid. Contains 100nM sodium azide and 0.7% BSA.
Catalog No. 50-200-9703