Learn More
New England Biolabs, Inc. BseRI – 1000 units
SDP
Catalog No. 502581804
Encompass
A restriction endonuclease that recognizes the sequence GAGGAGNNNNNNNN_NN^.
Catalog No. 50-258-1804 Supplier New England Biolabs, Inc. Supplier No. R0581L
A restriction endonuclease that recognizes the sequence GAGGAGNNNNNNNN_NN^.