Learn More
New England Biolabs, Inc. BseYI – 100 units
SDP
Catalog No. 50812384
Encompass
A restriction endonuclease that recognizes the sequence C^CCAG_C.
Catalog No. 50-812-384 Supplier New England Biolabs, Inc. Supplier No. R0635S
A restriction endonuclease that recognizes the sequence C^CCAG_C.