Learn More
New England Biolabs, Inc. BseYI – 500 units
SDP
Catalog No. 50812383
Encompass
A restriction endonuclease that recognizes the sequence C^CCAG_C.
Catalog No. 50-812-383 Supplier New England Biolabs, Inc. Supplier No. R0635L
A restriction endonuclease that recognizes the sequence C^CCAG_C.