Learn More
Research Products International Corp AEBSF, 1 G
SDP
Catalog No. 501977129
Encompass_Preferred
Specific and irreversible inhibitor of serine proteases. Inhibits chymotrypsin, kallikrein, plasmin, thrombin, trypsin and other related thrombolytic enzymes. Less toxic substitute for PMSF.
Catalog No. 50-197-7129 Supplier Research Products International Corp Supplier No. A200101.0
Add to Cart
Add to Cart
Specific and irreversible inhibitor of serine proteases. Inhibits chymotrypsin, kallikrein, plasmin, thrombin, trypsin and other related thrombolytic enzymes. Less toxic substitute for PMSF.