Learn More
Enzo Life Sciences AEBSF. hydrochloride (1g). CAS: 30827-99-7
SDP
Catalog No. 502008369
Encompass_Preferred
Serine protease inhibitor. Specific irreversible inhibitor of serine proteases including trypsin, chymotrypsin, plasmin, plasma kallikrein, and thrombin. Stable, non-toxic alternative to PMSF (ALX-270-184). Inhibitor of NAD(P)H oxidase. Activator of MAP kinases JNK and p38. Alternative name: 4-(2-Aminoethyl)benzenesulfonylfluoride . HCl, PefablocSC. Formula: C8H10FNO2S . HCl. MW: 239.7. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water or 100% ethanol. Long Term Storage: +4°C. Handling: Protect from moisture and light.
Catalog No. 50-200-8369 Supplier Enzo Life Sciences Supplier No. ALX270022G001
Add to Cart
Add to Cart
Serine protease inhibitor. Specific irreversible inhibitor of serine proteases including trypsin, chymotrypsin, plasmin, plasma kallikrein, and thrombin. Stable, non-toxic alternative to PMSF (ALX-270-184). Inhibitor of NAD(P)H oxidase. Activator of MAP kinases JNK and p38. Alternative name: 4-(2-Aminoethyl)benzenesulfonylfluoride . HCl, PefablocSC. Formula: C8H10FNO2S . HCl. MW: 239.7. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water or 100% ethanol. Long Term Storage: +4°C. Handling: Protect from moisture and light.