Learn More
Aviva Systems Biology Corp BSG Antibody - N-terminal region (ARP59754_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59754_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BSG. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human BSG. Alternate names: 5F7, CD147, EMMPRIN, M6, OK, TCSFApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-812