Learn More
New England Biolabs, Inc. BsgI – 250 units
SDP
Catalog No. 50812242
Encompass
A restriction endonuclease that recognizes the sequence GTGCAGNNNNNNNNNNNNNN_NN^.
Catalog No. 50-812-242 Supplier New England Biolabs, Inc. Supplier No. R0559L
A restriction endonuclease that recognizes the sequence GTGCAGNNNNNNNNNNNNNN_NN^.