Learn More
New England Biolabs, Inc. BsgI – 50 units
SDP
Catalog No. 50812243
Encompass
A restriction endonuclease that recognizes the sequence GTGCAGNNNNNNNNNNNNNN_NN^.
Catalog No. 50-812-243 Supplier New England Biolabs, Inc. Supplier No. R0559S
A restriction endonuclease that recognizes the sequence GTGCAGNNNNNNNNNNNNNN_NN^.