Learn More
Alkali Scientific BSH1001 One Block Digital Dry
SDP
Catalog No. 503029508
Encompass_Preferred
Traditional Dry Bath Lab Equipment Digital accuracy and convenience Single double and four block capacity Assorted blocks for a wide variety of tube sizes Custom blocks available upon request Our Alkali ScientificBSH1001 One Block Digital Dry Bath provides accurate inset and walk awayin digital temperature selection eliminating the need for external thermometers and repetitive infine tuningin of a temperature control knob Simply enter the desired temperature on the digital touchpad and the dry bath provides accurate temperature with real time monitoring that continuously maintains the selected temperature within 0 2C In addition a digital timer is built into the touchpad display Although it is not designed to shut off the heater it can be used to alert the user that a chosen time interval has timed out The cavities of the high grade aluminum blocks are precision machined to match the conical size and shape of 0 5 1 5 2 0 15 and 50ml tube sizes providing unsurpassed temperature trans
Catalog No. 50-302-9508 Supplier Alkali Scientific Supplier No. RS7149
Traditional Dry Bath Lab Equipment Digital accuracy and convenience Single double and four block capacity Assorted blocks for a wide variety of tube sizes Custom blocks available upon request Our Alkali ScientificBSH1001 One Block Digital Dry Bath provides accurate inset and walk awayin digital temperature selection eliminating the need for external thermometers and repetitive infine tuningin of a temperature control knob Simply enter the desired temperature on the digital touchpad and the dry bath provides accurate temperature with real time monitoring that continuously maintains the selected temperature within 0 2C In addition a digital timer is built into the touchpad display Although it is not designed to shut off the heater it can be used to alert the user that a chosen time interval has timed out The cavities of the high grade aluminum blocks are precision machined to match the conical size and shape of 0 5 1 5 2 0 15 and 50ml tube sizes providing unsurpassed temperature trans