Learn More
Alkali Scientific Alkali Scientific™ BSH1004 Four Block Digital Dry Bath, Must Purchase Blocks Separately, 115V
SDP
Catalog No. 503029524
Encompass_Preferred
Our Alkali Scientific™ BSH Digital Dry Baths provide accurate "set and walk away" digital temperature selection, eliminating the need for external thermometers and repetitive "fine tuning" of a temperature control knob. Simply enter the desired temperature on the digital touchpad and the Dry Bath provides accurate temperature with real time monitoring that continuously maintains the selected temperature within ±0.2°C. In addition, a digital timer is built into the touchpad display. Although it is not designed to shut off the heater, it can be used to alert the user that a chosen time interval has timed out. The cavities of the high grade aluminum blocks are precision machined to match the conical size and shape of 0.5, 1.5, 2.0, 15 and 50ml tube sizes, providing unsurpassed temperature transfer.Assorted blocks are available for compatibility with nearly all common laboratory tubes and plates. Custom blocks are also available upon request.
Catalog No. 50-302-9524 Supplier Alkali Scientific Supplier No. RS7151
Our Alkali Scientific™ BSH Digital Dry Baths provide accurate "set and walk away" digital temperature selection, eliminating the need for external thermometers and repetitive "fine tuning" of a temperature control knob. Simply enter the desired temperature on the digital touchpad and the Dry Bath provides accurate temperature with real time monitoring that continuously maintains the selected temperature within ±0.2°C. In addition, a digital timer is built into the touchpad display. Although it is not designed to shut off the heater, it can be used to alert the user that a chosen time interval has timed out. The cavities of the high grade aluminum blocks are precision machined to match the conical size and shape of 0.5, 1.5, 2.0, 15 and 50ml tube sizes, providing unsurpassed temperature transfer.Assorted blocks are available for compatibility with nearly all common laboratory tubes and plates. Custom blocks are also available upon request.