Learn More
Alkali Scientific Alkali Scientific™ BSH200-HL MyBlock Mini Dry Bath with Heated Lid
SDP
Catalog No. 503029540
Encompass_Preferred
Heated lid prevents evaporation & condensation Compact, fits in the palm of your hand Independent temperature control of lid & block Exchangeable blocks for 0.2 to 0.5ml tubes Room temperature to 37°C in just 2 minutes  The new Alkali Scientific™ MyBlock Mini BSH200-HL with heated lid fits almost anywhere and can even be used "on-the-go" in cars, boats or wherever a 12 volt power source is available. Its simple touch pad control with digital display is designed for "set and walk away" temperature selection and unrivaled accuracy. At less than 5 inches wide, The Alkali Scientific™ MyBlock Mini is truly the first personal block incubator
Catalog No. 50-302-9540 Supplier Alkali Scientific Supplier No. RS7135
Heated lid prevents evaporation & condensation Compact, fits in the palm of your hand Independent temperature control of lid & block Exchangeable blocks for 0.2 to 0.5ml tubes Room temperature to 37°C in just 2 minutes  The new Alkali Scientific™ MyBlock Mini BSH200-HL with heated lid fits almost anywhere and can even be used "on-the-go" in cars, boats or wherever a 12 volt power source is available. Its simple touch pad control with digital display is designed for "set and walk away" temperature selection and unrivaled accuracy. At less than 5 inches wide, The Alkali Scientific™ MyBlock Mini is truly the first personal block incubator