Learn More
Alkali Scientific Alkali Scientific™ BSH200 MyBlock Mini Dry Bath
SDP
Catalog No. 503029594
Encompass_Preferred
Compact, fits in the palm of your hand Exchangeable blocks for 0.2 to 50ml tubes Clear cover ensures temperature uniformity Room temperature to 37°C in just 2 minutes Portable; optional 12V power adapter The new Alkali Scientific™ BSH200 MyBlock Mini Dry Bathfits almost anywhere and can even be used "on-the-go" in cars, boats or wherever a 12 volt power source is available. Benchtop control of sample temperature has never been this easy or economical.Its simple touch pad control with digital display is designed for "set and walk away" temperature selection and unrivaled accuracy. At less than 5 inches wide, MyBlock Mini is truly the first personal block incubator.
Catalog No. 50-302-9594 Supplier Alkali Scientific Supplier No. RS7134
Compact, fits in the palm of your hand Exchangeable blocks for 0.2 to 50ml tubes Clear cover ensures temperature uniformity Room temperature to 37°C in just 2 minutes Portable; optional 12V power adapter The new Alkali Scientific™ BSH200 MyBlock Mini Dry Bathfits almost anywhere and can even be used "on-the-go" in cars, boats or wherever a 12 volt power source is available. Benchtop control of sample temperature has never been this easy or economical.Its simple touch pad control with digital display is designed for "set and walk away" temperature selection and unrivaled accuracy. At less than 5 inches wide, MyBlock Mini is truly the first personal block incubator.