Learn More
Alkali Scientific Alkali Scientific™ BSH300 MyBlock Mini Dry Bath with Heating and Cooling
SDP
Catalog No. 503029525
Encompass_Preferred
Provides accurate and uniform temperature control with powerful heating and cooling capabilities Compact, fits on any crowded benchtop Exchangeable blocks for 0.2 to 50ml tubes Simple touchpad, digital control Link two consecutive programs at once Blocks sold separately (please view the Accessories tab) The Alkali BSH300 MyBlock Mini Dry Bath with Cooling combines the features of compact construction, a broad temperature range, and multiple block options to make the perfect tool for laboratory incubations. The simple touch pad controls and digital display provide for “set and walk away” temperature selection and unrivaled accuracy. While perfect for bench use, the optional 12V adapter expands the utility of the Alkali MyBlock Mini to field use. At less than 4.3 inches wide, the MyBlock Mini is truly the first personal block incubator with cooling capabilities.
Catalog No. 50-302-9525 Supplier Alkali Scientific Supplier No. RS7136
Provides accurate and uniform temperature control with powerful heating and cooling capabilities Compact, fits on any crowded benchtop Exchangeable blocks for 0.2 to 50ml tubes Simple touchpad, digital control Link two consecutive programs at once Blocks sold separately (please view the Accessories tab) The Alkali BSH300 MyBlock Mini Dry Bath with Cooling combines the features of compact construction, a broad temperature range, and multiple block options to make the perfect tool for laboratory incubations. The simple touch pad controls and digital display provide for “set and walk away” temperature selection and unrivaled accuracy. While perfect for bench use, the optional 12V adapter expands the utility of the Alkali MyBlock Mini to field use. At less than 4.3 inches wide, the MyBlock Mini is truly the first personal block incubator with cooling capabilities.