Learn More
Alkali Scientific Alkali Scientific™ BSH5001-1B myBlock l- digital dry bath with 1 Quick-Flip blocks (BSWCMB) for tubes (0.2 to 2.0ml, PCR strips and PCR plate), 115V
SDP
Catalog No. 503029497
Encompass_Preferred
This Alkali Scientific™ BSH5001-1B myBlock l- digital dry bath features a unique Quick-flip™ block that accommodates 0.5 and 1.5/2.0 tubes on one side, and 0.2 tubes on the other! A powerful microprocessor gives the option for setting timed cycles or running continuously. An external temperature feedback probe can be placed in the sample tube to give you real-time monitoring and control of your sample's temperature. The Alkali Scientific™ MyBlock's see through lid provides better consistency in temperature and greatly lessens the formation of different temperature layers (stratification) by preserving a warm air jacket layer. The hinged lid can be removed with little effort or propped open to allow for larger tubes. This single block model  include our one of a kind Quick-Flip™ block. This new product has the ability to hold 0.5 to 2.0ml microcentrifuge tubes on one side or with a quick flip, they can hold 0.2ml tubes or PCR strips.
Catalog No. 50-302-9497 Supplier Alkali Scientific Supplier No. ASH50011B
This Alkali Scientific™ BSH5001-1B myBlock l- digital dry bath features a unique Quick-flip™ block that accommodates 0.5 and 1.5/2.0 tubes on one side, and 0.2 tubes on the other! A powerful microprocessor gives the option for setting timed cycles or running continuously. An external temperature feedback probe can be placed in the sample tube to give you real-time monitoring and control of your sample's temperature. The Alkali Scientific™ MyBlock's see through lid provides better consistency in temperature and greatly lessens the formation of different temperature layers (stratification) by preserving a warm air jacket layer. The hinged lid can be removed with little effort or propped open to allow for larger tubes. This single block model  include our one of a kind Quick-Flip™ block. This new product has the ability to hold 0.5 to 2.0ml microcentrifuge tubes on one side or with a quick flip, they can hold 0.2ml tubes or PCR strips.