Learn More
Alkali Scientific Alkali Scientific™ BSH5001 Alkali Scientific™ One-Block Digital Dry Bath 115V
SDP
Catalog No. 503029495
Encompass_Preferred
The MyBlock Series Dry Baths is a “next generation” product line that introduces several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls, timed or continuous operation and a removable hinged lid. In addition, an optional external temperature feedback probe can be positioned directly inside a sample tube, providing real-time control and monitoring of actual sample temperature. Digital temperature control (set and walk away) Hinged lid (improves uniformity & conserves energy) External (optional) temperature probe (measures actual sample temperature Available with Quick-Flip universal blocks (supports all common tube sizes, PCR strips and PCR plates) Patent Pending Blocks sold separately
Catalog No. 50-302-9495 Supplier Alkali Scientific Supplier No. ASH5001
The MyBlock Series Dry Baths is a “next generation” product line that introduces several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls, timed or continuous operation and a removable hinged lid. In addition, an optional external temperature feedback probe can be positioned directly inside a sample tube, providing real-time control and monitoring of actual sample temperature. Digital temperature control (set and walk away) Hinged lid (improves uniformity & conserves energy) External (optional) temperature probe (measures actual sample temperature Available with Quick-Flip universal blocks (supports all common tube sizes, PCR strips and PCR plates) Patent Pending Blocks sold separately