Learn More
New England Biolabs, Inc. BsiHKAI – 1000 units
SDP
Catalog No. 50812264
Encompass
A restriction endonuclease that recognizes the sequence G_WGCW^C.
Catalog No. 50-812-264 Supplier New England Biolabs, Inc. Supplier No. R0570S
A restriction endonuclease that recognizes the sequence G_WGCW^C.