Learn More
New England Biolabs, Inc. BsiWI – 1500 units
SDP
Catalog No. 50812230
Encompass
A restriction endonuclease that recognizes the sequence C^GTAC_G.
Catalog No. 50-812-230 Supplier New England Biolabs, Inc. Supplier No. R0553L
A restriction endonuclease that recognizes the sequence C^GTAC_G.