Learn More
New England Biolabs, Inc. BsiWI – 300 units
SDP
Catalog No. 50812231
Encompass
A restriction endonuclease that recognizes the sequence C^GTAC_G.
Catalog No. 50-812-231 Supplier New England Biolabs, Inc. Supplier No. R0553S
A restriction endonuclease that recognizes the sequence C^GTAC_G.