Learn More
Medchemexpress LLC BSJ-02-162 | 2139329-47-6 | 98.1% | C43H49N11O7 | 1 ML
SDP
Catalog No. 5000356342
Encompass_Preferred
BSJ-02-162, also known as CDK4/6-IN-11, is a potent PROTAC CDK4/6 degrader. It is specifically designed with a thalidomide-based E3 ligase ligand, Palbociclib as a target protein ligand, and a PROTAC linker, making it suitable for targeted protein degradation studies.
  • Potent PROTAC CDK4/6 degrader
  • Composed of a thalidomide-based E3 ligase ligand
  • Includes a target protein ligand
  • Features a PROTAC linker
  • Available in solid and solution formats
  • Soluble in DMSO
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-56342 Supplier Medchemexpress LLC Supplier No. HY14499510MM/1ML
BSJ-02-162, also known as CDK4/6-IN-11, is a potent PROTAC CDK4/6 degrader. It is specifically designed with a thalidomide-based E3 ligase ligand, Palbociclib as a target protein ligand, and a PROTAC linker, making it suitable for targeted protein degradation studies.
  • Potent PROTAC CDK4/6 degrader
  • Composed of a thalidomide-based E3 ligase ligand
  • Includes a target protein ligand
  • Features a PROTAC linker
  • Available in solid and solution formats
  • Soluble in DMSO
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month