Learn More
Medchemexpress LLC BSJ-02-162 (CDK4/6-IN-11) | 2139329-47-6 | 98.1% | 831.92 g/mol | C43H49N11O7 | 5 MG
SDP
Catalog No. 5000221594
Encompass_Preferred
BSJ-02-162 is a proteolysis-targeting chimera (PROTAC) small molecule for research use that induces degradation of cyclin-dependent kinases CDK4 and CDK6. It links an E3 ligase ligand to a CDK4/6-targeting ligand via a chemical linker to promote targeted proteasomal degradation in biochemical and cell-based studies. Not for human or clinical use.
  • Induces selective degradation of CDK4 and CDK6 in cell-based assays.
  • Suitable for biochemical and cell-based research into cell cycle pathways.
  • Available as solid powder and as a DMSO solution for flexible dosing.
  • High reported purity supports reproducible experimental results.
  • Compatible with common assay formats and routine handling.
Catalog No. 50-002-21594 Supplier Medchemexpress LLC Supplier No. HY1449955MG
BSJ-02-162 is a proteolysis-targeting chimera (PROTAC) small molecule for research use that induces degradation of cyclin-dependent kinases CDK4 and CDK6. It links an E3 ligase ligand to a CDK4/6-targeting ligand via a chemical linker to promote targeted proteasomal degradation in biochemical and cell-based studies. Not for human or clinical use.
  • Induces selective degradation of CDK4 and CDK6 in cell-based assays.
  • Suitable for biochemical and cell-based research into cell cycle pathways.
  • Available as solid powder and as a DMSO solution for flexible dosing.
  • High reported purity supports reproducible experimental results.
  • Compatible with common assay formats and routine handling.