Learn More
Medchemexpress LLC Bsj-02-162 (cdk4/6-in-11) | 2139329-47-6 | 98.1% | C43H49N11O7 | 25 MG
SDP
Catalog No. 5000357138
Encompass_Preferred
CDK4/6-IN-11 is a potent PROTAC CDK4/6 degrader. It is composed of a thalidomide-based E3 ligase ligand, a target protein ligand (Palbociclib), and a PROTAC linker. This product is intended for research use only.
  • Potent PROTAC CDK4/6 degrader
  • Comprised of a thalidomide-based E3 ligase ligand
  • Includes a target protein ligand (Palbociclib)
  • Incorporates a PROTAC linker
  • E3 ligase ligand and linker can form Thalidomide-NH-amido-C4-Br
Catalog No. 50-003-57138 Supplier Medchemexpress LLC Supplier No. 25MGHY144995
CDK4/6-IN-11 is a potent PROTAC CDK4/6 degrader. It is composed of a thalidomide-based E3 ligase ligand, a target protein ligand (Palbociclib), and a PROTAC linker. This product is intended for research use only.
  • Potent PROTAC CDK4/6 degrader
  • Comprised of a thalidomide-based E3 ligase ligand
  • Includes a target protein ligand (Palbociclib)
  • Incorporates a PROTAC linker
  • E3 ligase ligand and linker can form Thalidomide-NH-amido-C4-Br