Learn More
Medchemexpress LLC BSJ-02-162 (CDK4/6-IN-11) | 2139329-47-6 | 98.1% | C43H49N11O7 | 50 MG
SDP
Catalog No. 5000354439
Encompass_Preferred
CDK4/6-IN-11 is a potent PROTAC CDK4/6 degrader. It is composed of a thalidomide-based E3 ligase ligand, a target protein ligand (Palbociclib), and a PROTAC linker. The E3 ligase ligand and linker can combine to form Thalidomide-NH-amido-C4-Br. The product is intended for research use only.
  • Potent PROTAC CDK4/6 degrader, effectively degrades CDK4/6.
  • Composed of well-defined components including a thalidomide-based E3 ligase ligand, Palbociclib as a target protein ligand, and a PROTAC linker.
  • High purity of 98.07% ensures reliable experimental results.
  • Available in various quantities, ranging from 5 mg to 500 mg, catering to different research needs.
  • Detailed documentation including Data Sheet, SDS, COA, HNMR, RP-HPLC, MS, and Handling Instructions are available.
Catalog No. 50-003-54439 Supplier Medchemexpress LLC Supplier No. HY14499550MG
CDK4/6-IN-11 is a potent PROTAC CDK4/6 degrader. It is composed of a thalidomide-based E3 ligase ligand, a target protein ligand (Palbociclib), and a PROTAC linker. The E3 ligase ligand and linker can combine to form Thalidomide-NH-amido-C4-Br. The product is intended for research use only.
  • Potent PROTAC CDK4/6 degrader, effectively degrades CDK4/6.
  • Composed of well-defined components including a thalidomide-based E3 ligase ligand, Palbociclib as a target protein ligand, and a PROTAC linker.
  • High purity of 98.07% ensures reliable experimental results.
  • Available in various quantities, ranging from 5 mg to 500 mg, catering to different research needs.
  • Detailed documentation including Data Sheet, SDS, COA, HNMR, RP-HPLC, MS, and Handling Instructions are available.