Learn More
Medchemexpress LLC BSJ-03-123 | 2361493-16-3 | 99.19% | 937.01 | 10 MG
SDP
Catalog No. 5000347174
Encompass_Preferred
BSJ-03-123 is a PROTAC connected by ligands for Cereblon and CDK, functioning as a potent and novel CDK6-selective small-molecule degrader. It causes a pronounced anti-proliferative effect in CDK6-dependent AML cell lines by inducing a G1 cell-cycle arrest without a measurable increase in apoptosis.
  • Potent and novel CDK6-selective small-molecule degrader
  • Induces G1 cell-cycle arrest in AML cell lines
  • Causes anti-proliferative effect in CDK6-dependent AML cell lines
  • Does not significantly increase apoptosis
Catalog No. 50-003-47174 Supplier Medchemexpress LLC Supplier No. HY111556100MG
BSJ-03-123 is a PROTAC connected by ligands for Cereblon and CDK, functioning as a potent and novel CDK6-selective small-molecule degrader. It causes a pronounced anti-proliferative effect in CDK6-dependent AML cell lines by inducing a G1 cell-cycle arrest without a measurable increase in apoptosis.
  • Potent and novel CDK6-selective small-molecule degrader
  • Induces G1 cell-cycle arrest in AML cell lines
  • Causes anti-proliferative effect in CDK6-dependent AML cell lines
  • Does not significantly increase apoptosis