Learn More
Medchemexpress LLC HY-111556 5mg Medchemexpress, BSJ-03-123 CAS: Purity:>98%
SDP
Catalog No. 501932375
Encompass_Preferred
Medchemexpress, HY-111556 5mg BSJ-03-123 CAS: BSJ-03-123 is a potent and novel CDK6 -selective small-molecule degrader (PROTAC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2375 Supplier Medchemexpress LLC Supplier No. HY1115565MG
Add to Cart
Add to Cart
Medchemexpress, HY-111556 5mg BSJ-03-123 CAS: BSJ-03-123 is a potent and novel CDK6 -selective small-molecule degrader (PROTAC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.