Learn More
Medchemexpress LLC Bsj-03-204 | 2349356-09-6 | 99.76% | 832.90 | 100 MG
SDP
Catalog No. 5000348702
Encompass_Preferred
BSJ-03-204 is a PROTAC (proteolysis-targeting chimera) that connects ligands for Cereblon and CDK. It is a potent and selective Palbociclib-based CDK4/6 dual degrader, demonstrating anti-cancer activity and potent anti-proliferative effects on MCL cell lines.
  • Potent and selective Palbociclib-based CDK4/6 dual degrader
  • Does not induce IKZF1/3 degradation
  • Exhibits anti-cancer activity
  • Potent anti-proliferative effects on MCL cell lines
  • Induces a G1 arrest
Catalog No. 50-003-48702 Supplier Medchemexpress LLC Supplier No. HY136250100MG
BSJ-03-204 is a PROTAC (proteolysis-targeting chimera) that connects ligands for Cereblon and CDK. It is a potent and selective Palbociclib-based CDK4/6 dual degrader, demonstrating anti-cancer activity and potent anti-proliferative effects on MCL cell lines.
  • Potent and selective Palbociclib-based CDK4/6 dual degrader
  • Does not induce IKZF1/3 degradation
  • Exhibits anti-cancer activity
  • Potent anti-proliferative effects on MCL cell lines
  • Induces a G1 arrest