Learn More
Medchemexpress LLC BSJ-03-204 (triTFA) | 99.9% | 1174.97 | 25 MG
SDP
Catalog No. 5000347852
Encompass_Preferred
BSJ-03-204 triTFA is a PROTAC connected by ligands for Cereblon and CDK. It is a potent and selective Palbociclib-based CDK4/6 dual degrader, with IC50s of 26.9 nM for CDK4/D1 and 10.4 nM for CDK6/D1. This compound does not induce IKZF1/3 degradation and demonstrates anti-cancer activity.
  • PROTAC connected by ligands for Cereblon and CDK.
  • Potent and selective Palbociclib-based CDK4/6 dual degrader.
  • Demonstrates potent anti-proliferative effects on MCL cell lines.
  • Induces a G1 arrest.
  • Selectively degrades CDK4/6 in WT cells without affecting IKZF1/3 degradation.
  • Exhibits anti-cancer activity.
Catalog No. 50-003-47852 Supplier Medchemexpress LLC Supplier No. 25MGHY136250A
BSJ-03-204 triTFA is a PROTAC connected by ligands for Cereblon and CDK. It is a potent and selective Palbociclib-based CDK4/6 dual degrader, with IC50s of 26.9 nM for CDK4/D1 and 10.4 nM for CDK6/D1. This compound does not induce IKZF1/3 degradation and demonstrates anti-cancer activity.
  • PROTAC connected by ligands for Cereblon and CDK.
  • Potent and selective Palbociclib-based CDK4/6 dual degrader.
  • Demonstrates potent anti-proliferative effects on MCL cell lines.
  • Induces a G1 arrest.
  • Selectively degrades CDK4/6 in WT cells without affecting IKZF1/3 degradation.
  • Exhibits anti-cancer activity.