Learn More
TARGETMOL CHEMICALS INC BSJ-04-122 10MG
SDP
Catalog No. 502576328
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. BSJ-04-122 is a covalent MKK4/7 dual inhibitor that inhibits MKK4 and MKK7 with IC50 values of 4 nM and 181 nM respectively.BSJ-04-122 can be used for cancer research. purity: 97%
Catalog No. 50-257-6328 Supplier TARGETMOL CHEMICALS INC Supplier No. T7318210MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. BSJ-04-122 is a covalent MKK4/7 dual inhibitor that inhibits MKK4 and MKK7 with IC50 values of 4 nM and 181 nM respectively.BSJ-04-122 can be used for cancer research. purity: 97%