Learn More
Medchemexpress LLC BSJ-04-122 | 2513289-74-0 | C15H12ClN5O | 1 ML
SDP
Catalog No. 5000373201
Encompass_Preferred
BSJ-04-122 is a covalent MKK4/7 dual inhibitor. It inhibits MKK4 and MKK7 with IC50 values of 4 nM and 181 nM, respectively. It can be used for cancer research.
  • Covalent MKK4/7 dual inhibitor
  • Inhibits MKK4 with IC50 of 4 nM
  • Inhibits MKK7 with IC50 of 181 nM
  • Useful for cancer research
Catalog No. 50-003-73201 Supplier Medchemexpress LLC Supplier No. HY15218510MM/1ML
BSJ-04-122 is a covalent MKK4/7 dual inhibitor. It inhibits MKK4 and MKK7 with IC50 values of 4 nM and 181 nM, respectively. It can be used for cancer research.
  • Covalent MKK4/7 dual inhibitor
  • Inhibits MKK4 with IC50 of 4 nM
  • Inhibits MKK7 with IC50 of 181 nM
  • Useful for cancer research