Learn More
Medchemexpress LLC BSJ-04-122 | 2513289-74-0 | C15H12ClN5O | 5 MG
SDP
Catalog No. 5000373418
Encompass_Preferred
BSJ-04-122 is a highly potent covalent dual inhibitor targeting MKK4 and MKK7, key kinases involved in cellular signaling pathways. It demonstrates strong inhibitory activity against MKK4 with an IC50 value of 4 nM and against MKK7 with an IC50 value of 181 nM. This compound is primarily utilized in cancer research, exhibiting its ability to induce a robust reduction in JNK phosphorylation.
  • Potent covalent dual inhibitor of MKK4 and MKK7
  • Inhibits MKK4 with an IC50 of 4 nM
  • Inhibits MKK7 with an IC50 of 181 nM
  • Induces robust reduction in JNK phosphorylation
  • Enhances antiproliferative effects when combined with JNK-IN-8
  • Useful for cancer research applications
Catalog No. 50-003-73418 Supplier Medchemexpress LLC Supplier No. HY1521855MG
BSJ-04-122 is a highly potent covalent dual inhibitor targeting MKK4 and MKK7, key kinases involved in cellular signaling pathways. It demonstrates strong inhibitory activity against MKK4 with an IC50 value of 4 nM and against MKK7 with an IC50 value of 181 nM. This compound is primarily utilized in cancer research, exhibiting its ability to induce a robust reduction in JNK phosphorylation.
  • Potent covalent dual inhibitor of MKK4 and MKK7
  • Inhibits MKK4 with an IC50 of 4 nM
  • Inhibits MKK7 with an IC50 of 181 nM
  • Induces robust reduction in JNK phosphorylation
  • Enhances antiproliferative effects when combined with JNK-IN-8
  • Useful for cancer research applications