Learn More
TARGETMOL CHEMICALS INC BSJ-4-116 5MG
SDP
Catalog No. 502576211
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BSJ-4-116 is a highly potent and selective CDK12 degrader (PROTAC) with an IC50 of 6 nM. It downregulates DDR genes through a premature termination of transcription primarily through increasing poly(adenylation). purity: 99%
Catalog No. 50-257-6211 Supplier TARGETMOL CHEMICALS INC Supplier No. T91175MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BSJ-4-116 is a highly potent and selective CDK12 degrader (PROTAC) with an IC50 of 6 nM. It downregulates DDR genes through a premature termination of transcription primarily through increasing poly(adenylation). purity: 99%