Learn More
Medchemexpress LLC HY-139039 5mg , BSJ-4-116 CAS:2519823-34-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1390395MG
Encompass_Preferred
HY-139039 5mg BSJ-4-116 CAS: 2519823-34-6 BSJ-4-116 is a PROTAC connected by ligands for Cereblon and CDK. BSJ-4-116 is a highly potent and selective CDK12 degrader (PROTAC) with an IC50 of 6 nM. BSJ-4-116 downregulates DDR genes through a premature termination of transcription, primarily through increasing poly(adenylation). BSJ-4-116 exhibits potent antiproliferative effects, alone and in combination with the poly(ADP-ribose) polymerase inhibitor Olaparib (HY-10162). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9511