Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix (BSL-3) Heartland Virus (Strain: MO4) Culture Fluid
SDP

Catalog No. 22158315 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-315 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-315 Supplier ZeptoMetrix Supplier No. 0810576CF
Add to Cart
Add to Cart

Heartland Virus (Strain: MO4) is a negative-sense single-stranded RNA virus from the Bunyaviridae family.

  • This product is intended for research, analytical testing during product development, or quality assurance testing. Viral culture fluids are sold as consumable testing materials, and are not for propagation or commercialization.
  • Applications include: Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays.
  • Quality Control

Specifications

Content And Storage Viral culture fluids should be stored at -65°C or below.
Description Repetitive freezing and thawing is not recommended (aliquot material if necessary). Titer will be altered by multiple freeze-thaws.
Product Type Culture Fluid
For Use With (Application) Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays
For Use With (Equipment) To avoid cross-contamination, use separate pipette tips for all reagents.
Includes Each frozen aliquot contains 1 mL of titered viral culture fluid.
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.