Learn More
New England Biolabs, Inc. BslI – 1000 units
SDP
Catalog No. 50812235
Encompass
A restriction endonuclease that recognizes the sequence CCNN_NNN^NNGG.
Catalog No. 50-812-235 Supplier New England Biolabs, Inc. Supplier No. R0555S
A restriction endonuclease that recognizes the sequence CCNN_NNN^NNGG.