Learn More
New England Biolabs, Inc. BslI – 5000 units
SDP
Catalog No. 50812234
Encompass
A restriction endonuclease that recognizes the sequence CCNN_NNN^NNGG.
Catalog No. 50-812-234 Supplier New England Biolabs, Inc. Supplier No. R0555L
A restriction endonuclease that recognizes the sequence CCNN_NNN^NNGG.