Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Bsm DNA Polymerase, large fragment (8 U/μL)

Catalog No. FEREP0691 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1,600 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREP0691 1,600 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREP0691 Supplier Thermo Scientific™ Supplier No. EP0691
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific Bsm DNA Polymerase, Large Fragment, is equivalent to Bst DNA polymerase, which catalyzes 5' → 3' synthesis of DNA and lacks 5' → 3' and 3' → 5' exonuclease activities.

Thermo Scientific Bsm DNA Polymerase, Large Fragment, is equivalent to Bst DNA polymerase, which catalyzes 5' → 3' synthesis of DNA and lacks 5' → 3' and 3' → 5' exonuclease activities. Sourced from E. coli cells with a cloned polA gene from Bacillus smithii, Bsm DNA Polymerase, Large Fragment, features strong strand displacement activity and is active in a wide range of temperatures (30–63°C), with an optimum of activity at 60°C. With high functional similarity to Bst DNA Polymerase, Large Fragment, this enzyme can serve as alternative in most applications including loop-mediated isothermal amplification (LAMP).

Thermo Scientific Bsm DNA Polymerase, Large Fragment, is equivalent to Bst DNA polymerase, which catalyzes 5' → 3' synthesis of DNA and lacks 5' → 3' and 3' → 5' exonuclease activities. Sourced from E. coli cells with a cloned polA gene from Bacillus smithii, Bsm DNA Polymerase, Large Fragment, features strong strand displacement activity and is active in a wide range of temperatures (30–63°C), with an optimum of activity at 60°C. With high functional similarity to Bst DNA Polymerase, Large Fragment, this enzyme can serve as an alternative in most applications, including loop-mediated isothermal amplification (LAMP).

Applications
Bsm DNA Polymerase, Large Fragment, may be used in a wide variety of applications including:
• Loop-mediated isothermal amplification (LAMP)
• Whole genome amplification (WGA)
• Ramification amplification (RAM)
• Random-primed DNA labeling
• Labeling by fill-in 5'-overhangs of dsDNA

TRUSTED_SUSTAINABILITY

Specifications

Product Type Stand-alone enzyme
For Use With (Application) LAMP, RT-LAMP, WGA, RAM
Polymerase Bsm DNA Polymerase
Quantity 1,600 units
Content And Storage

• Bsm DNA Polymerase, Large Fragment (200 μL at 8 U/μL)
• 10X Bsm Buffer (1.25 mL)

Store at –15 to –25°C.

Optimal Reaction Temperature 60°C
Shipping Condition Dry Ice
Concentration 8 U/μL
Reaction Time 30 to 60 minutes
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.