Learn More
New England Biolabs, Inc. BsmAI – 1000 units
SDP
Catalog No. 50812190
Encompass
A restriction endonuclease that recognizes the sequence GTCTCN^NNNN_.
Catalog No. 50-812-190 Supplier New England Biolabs, Inc. Supplier No. R0529S
A restriction endonuclease that recognizes the sequence GTCTCN^NNNN_.