Learn More
New England Biolabs, Inc. BsmAI – 5000 units
SDP
Catalog No. 50812189
Encompass
A restriction endonuclease that recognizes the sequence GTCTCN^NNNN_.
Catalog No. 50-812-189 Supplier New England Biolabs, Inc. Supplier No. R0529L
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
A restriction endonuclease that recognizes the sequence GTCTCN^NNNN_.