Learn More
New England Biolabs, Inc. BsmBI-v2 – 200 units
SDP
Catalog No. NC1807712
Encompass
A Type IIS restriction endonuclease that recognizes the sequence CGTCTCN^NNNN and is optimized for Golden Gate Assembly.
  • Time-Saver qualified for digestion in 5-15 minutes
  • Used in Golden Gate Assembly
  • Type IIS Res recognize asymmetric DNA sequences; cleave outside of their recognition site
  • Supplied with 1 vial of Gel Loading Dye, Purple (6X)
Catalog No. NC1807712 Supplier New England Biolabs, Inc. Supplier No. R0739S
A Type IIS restriction endonuclease that recognizes the sequence CGTCTCN^NNNN and is optimized for Golden Gate Assembly.
  • Time-Saver qualified for digestion in 5-15 minutes
  • Used in Golden Gate Assembly
  • Type IIS Res recognize asymmetric DNA sequences; cleave outside of their recognition site
  • Supplied with 1 vial of Gel Loading Dye, Purple (6X)