Learn More
New England Biolabs, Inc. BsmFI – 100 units
SDP
Catalog No. 50812268
Encompass
A restriction endonuclease that recognizes the sequence GGGACNNNNNNNNNN^NNNN_.
Catalog No. 50-812-268 Supplier New England Biolabs, Inc. Supplier No. R0572S
A restriction endonuclease that recognizes the sequence GGGACNNNNNNNNNN^NNNN_.