Learn More
New England Biolabs, Inc. BsmI – 2500 units
SDP
Catalog No. 50811990
Encompass
A restriction endonuclease that recognizes the sequence GAATG_CN^.
Catalog No. 50-811-990 Supplier New England Biolabs, Inc. Supplier No. R0134L
A restriction endonuclease that recognizes the sequence GAATG_CN^.