Learn More
Aviva Systems Biology Corp BSND Antibody - middle region (ARP59578_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59578_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BSND. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human BSND. Alternate names: BART, DFNB73, MGC119283, MGC119284, MGC119285ApplicationWB. Species reactivity: Human
Catalog No. 50-286-747