Learn More
Selleck Chemical LLC BSO -sulfoximine 5mg
SDP
Supplier:  Selleck Chemical LLC S97285MG
Encompass_Preferred
BSO (L-Buthionine-(S,R)-sulfoximine, L-Buthionine sulfoximine) is a cell-permeable, potent, fast acting and irreversible inhibitor ofg-glutamylcysteine synthetase (?;-glutamylcysteine synthetase, ?;-GCS) and depletes cellular glutathione levels. The IC50 of BSO on melanoma, breast and ovarian tumor specimens are 1.9 ?M, 8.6 ?M, and 29 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3504